Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Phosphate (Mono- and Di- Ester mixture), TCI America™
SDP

Catalog No. I024725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I024725ML 25 mL
I0247500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I024725ML Supplier TCI America Supplier No. I024725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52933-00-3
Molecular Formula C9H24O8P2
Molecular Weight (g/mol) 322.23
MDL Number MFCD00135965
InChI Key RTIXFJOJNSXSOB-UHFFFAOYSA-N
Synonym phosphoric acid isopropyl ester, isopropyl phosphate mono-and di-ester mixture
PubChem CID 44630356

Specifications

Color Yellow
Quantity 25 mL
UN Number 1793
Formula Weight 182.16
Physical Form Liquid
Chemical Name or Material Isopropyl Phosphate (Mono- and Di- Ester mixture)
EINECSNumber (2)-1981&(2)-1986
RTECSNumber TC6320000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.