Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
75-31-0
C3H9N
59.11
JJWLVOIRVHMVIS-UHFFFAOYSA-N
isopropylamine,2-propanamine,2-aminopropane,monoisopropylamine,2-propylamine,sec-propylamine,1-methylethylamine,isopropyl amine,isopropilamina,propane, 2-amino
6363
CHEBI:15739
propan-2-amine
CC(C)N
75-31-0
C3H9N
59.11
JJWLVOIRVHMVIS-UHFFFAOYSA-N
isopropylamine,2-propanamine,2-aminopropane,monoisopropylamine,2-propylamine,sec-propylamine,1-methylethylamine,isopropyl amine,isopropilamina,propane, 2-amino
6363
CHEBI:15739
propan-2-amine
CC(C)N
Viscosity 0.3 mPa.s (20°C)
Quantity 1L
Water 0.5% Max. (K.F.)
Specific Gravity 0.69
Density 0.6900g/mL
Color White
Flash Point −37°C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 33.0°C to 34.0°C
Refractive Index 1.3725 to 1.3755
Melting Point -101.0°C
Assay Percent Range 99%
Linear Formula (CH3)2CHNH2
Beilstein 04,152
Fieser 13,164
Merck Index 15,5255
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 59.11
Physical Form Crystalline Powder, Needles or Crystals
Percent Purity 99%
Chemical Name or Material Isopropylamine
Show More Show Less

GHS P Statement May cause respiratory irritation. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger