Learn More
Isopropylidene malonate, 97%, Thermo Scientific Chemicals
Description
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Isopropylidene malonate is an important reagent in synthetic chemistry and involved in the formation of new C-C bonds, rings, amides, esters, and acids. The presence of carbonyl groups at C-4 and C-6 positions favors the alkylation and acylation reactions due to its acidic nature. It is also employed as an antimicrobial agent. It is used in the preparation of 4-pyridyl-substituted heterocycles, 2-substituted indoles, isofraxidin and macrocyclic beta-keto lactones. Further, it is involved in the Knoevenagel condensation reaction with aldehydes.
Solubility
Soluble in water. Slightly soluble in ethanol and chloroform.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 92°C to 96°C |
Quantity | 25 g |
Beilstein | 117310 |
Sensitivity | Moisture sensitive |
Merck Index | 14,5817 |
Solubility Information | Soluble in water. Slightly soluble in ethanol and chloroform. |
Formula Weight | 144.13 |
Percent Purity | 98% |
Chemical Name or Material | Isopropylidene malonate |
Safety and Handling
Hazard Category | H315 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 217-992-8 |
RTECSNumber | JH0800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.