Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylmagnesium chloride - Lithium chloride complex, 1.3M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
745038-86-2,109-99-9
C3H7Cl2LiMg
145.234
MFCD07784514
DBTNVRCCIDISMV-UHFFFAOYSA-L
turbo grignard,iprmgcl licl,i-prmgcl licl,i-prmgcl.licl,isopropylmagnesiumchloride licl,isopropylmagnesium chloride licl,isopropyl magnesium chloride licl,cwtureabailgik-uhfffaoysa-l,isopropylmagnesium lithium chloride,isopropyl magnesium chloride li-cl
11275082
lithium;magnesium;propane;dichloride
[Li+].C[CH-]C.[Mg+2].[Cl-].[Cl-]
745038-86-2,109-99-9
C3H7Cl2LiMg
145.234
MFCD07784514
DBTNVRCCIDISMV-UHFFFAOYSA-L
turbo grignard,iprmgcl licl,i-prmgcl licl,i-prmgcl.licl,isopropylmagnesiumchloride licl,isopropylmagnesium chloride licl,isopropyl magnesium chloride licl,cwtureabailgik-uhfffaoysa-l,isopropylmagnesium lithium chloride,isopropyl magnesium chloride li-cl
11275082
lithium;magnesium;propane;dichloride
[Li+].C[CH-]C.[Mg+2].[Cl-].[Cl-]
Concentration 1.1 to 1.5M
Density 0.9800g/mL
Formula Weight 145.24
Physical Form Solution
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.98
Storage Information Product may develop some turbidity or precipitate
Color Gray to Yellow
Quantity 100mL
Flash Point −21°C
Chemical Name or Material Isopropylmagnesium chloride - Lithium chloride complex, 14% solution in THF
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. Highly flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection.

Danger