Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopulegol, mixture of isomers, 97%

Catalog No. AAB2026009
Click to view available options
Quantity:
10 g

CAS: 7786-67-6 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-67-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00134655
InChI Key ZYTMANIQRDEHIO-UHFFFAOYNA-N
Synonym p-menth-8-en-3-ol, isopulegol, 8 9-p-menthen-3-ol, 5-methyl-2-prop-1-en-2-yl cyclohexanol, cyclohexanol, 5-methyl-2-1-methylethenyl, 1-methyl-4-isopropenylcyclohexan-3-ol, dl-isopulegol, p-8 9-menthen-3-ol, 5-methyl-2-1-methylvinyl cyclohexan-1-ol, 5-methyl-2-prop-1-en-2-yl cyclohexan-1-ol
PubChem CID 24585
IUPAC Name 5-methyl-2-prop-1-en-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(O)C1)C(C)=C

Specifications

Density 0.91
Boiling Point 90°C to 92°C (12 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.4725
Quantity 10 g
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material Isopulegol, mixture of isomers
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
EINECSNumber 232-102-8
RTECSNumber OT0190000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.