Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-5-sulfonic Acid 96.0+%, TCI America™
SDP

Catalog No. I0470250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0470250G 250 g
I047025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0470250G Supplier TCI America Supplier No. I0470250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27655-40-9
Molecular Formula C9H7NO3S
Molecular Weight (g/mol) 209.219
MDL Number MFCD00134089
InChI Key YFMJTLUPSMCTOQ-UHFFFAOYSA-N
Synonym 5-isoquinolinesulfonic acid, 5-sulfoisoquinoline, 5-isoquinolinesulfonicacid, 5-sulphoisoquinoline, pubchem23533, acmc-1chvq, isoquinoline-5-sulphonic acid, ksc203s9f
PubChem CID 241599
IUPAC Name isoquinoline-5-sulfonic acid
SMILES C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)O

Specifications

Color White-Yellow
Quantity 250 g
UN Number 2585
Formula Weight 209.22
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-5-sulfonic Acid
EINECSNumber (5)-0826
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.