Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline, 97%

Catalog No. p-152147
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC325031000

Supplier: Thermo Scientific Chemicals 325031000

Only null left
Add to Cart

CAS: 119-65-3 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-65-3
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006898,MFCD31699977
InChI Key AWJUIBRHMBBTKR-UHFFFAOYSA-N
Synonym 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech
PubChem CID 8405
ChEBI CHEBI:16092
IUPAC Name isoquinoline
SMILES C1=CC=C2C=NC=CC2=C1

Specifications

Melting Point 24°C to 28°C
Color Brown
Density 1.0900g/mL
Boiling Point 242°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.623
Quantity 100 g
Beilstein 20, 380
Specific Gravity 1.09
Merck Index 15, 5268
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted acids,misible with many organic solvents
Viscosity 3.25 mPa.s
Formula Weight 129.16
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material Isoquinoline, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
Do not eat,drink or smoke when using this product.
IF ON SKIN: Wash with plenty
EINECSNumber 204-341-8
RTECSNumber NW6825000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.