Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isosorbide, 98%

Catalog No. AC351911000
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

CAS: 652-67-5 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-67-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00064827
InChI Key KLDXJTOLSGUMSJ-JGWLITMVSA-N
Synonym isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol
PubChem CID 12597
IUPAC Name (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol
SMILES C1C(C2C(O1)C(CO2)O)O

Specifications

Melting Point 56.0°C to 62.0°C
Color Beige or White to Yellow
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystals, Crystalline Solid or Mass
Assay Percent Range 97.5% min. (GC)
Merck Index 15, 5270
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material Isosorbide
Show More Show Less
EINECSNumber 211-492-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.