Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators

Catalog No. fbg30cpga Shop All Fisherbrand Products
Click to view available options
Capacity (English):
27 cu. ft.
49 cu. ft.
72 cu. ft.
Shelves:
12 Full, 2 Half
4 Full, 3 Half
8 Full, 2 Half
Color:
Gray
White
Interior:
Painted White Steel
Stainless Steel
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators are designed with your needs in mind.

  • Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
  • Internal plug/outlet
  • Special safety sensor turns the internal plug/outlet off if a hydrocarbon (coolant) leak is detected
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 27 cu. ft.
Capacity (Metric) 764 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) 5°C
Refrigerant R290
Door Style Glass
No. of Doors 1
Shelves 4 Full, 3 Half
Capacity (English) Shelf 100 lb./full shelf; 50 lb./half shelf
Capacity (Metric) Shelf 45 kg/full shelf, 22.5 kg/half shelf
Interior Painted White Steel
Certifications/Compliance cULus, ENERGYSTAR
Color White
Control Type Microprocessor
Ports 1 Rear access port and 2 side access ports
Warranty 2 years parts and labor, plus 3 year compressor warranty
Model CPGA
Access Security Key Lock
Dimensions (D x W x H) Interior 28.5 x 27 x 61.75 in. (724 x 686 x 1569 mm)
Dimensions (L x W x H) Exterior 32 x 30.75 x 81 in.(813 x 781 x 813 mm)
Amperage 3 A
Plug Type NEMA 5-15 P
Electrical Requirements 115 V 60 Hz
Shipping Weight (English) 386 lb.
Shipping Weight (Metric) 175 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.