Learn More
Pyramex™ Isotope™ Safety Eyewear
Dual frame design features a rubber gasket for a comfortable fit while protecting from splash and dust
$121.10 - $558.00
Specifications
Certifications/Compliance ANSI Z87.1 ,CSA Z94.3-07
Description Dielectric
Certifications/Compliance (Europe) EN-166
Frame Color Black
Weight (Metric) 41 g
View More Specs
Products 4
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-155-349
View Documents
Pyramex™
GB9410STM
Clear
Case of 12 for $558.00
Only null left
Edge
Add to Cart
19-155-350
View Documents
Pyramex™
GB9420STM
Gray
Case of 12 for $533.00
Only null left
Add to Cart
19-155-351
View Documents
Pyramex™
GB9430STM
Amber
Case of 12 for $533.00
Only null left
Add to Cart
19-155-352
View Documents
Pyramex™
GB9480ST
Indoor/Outdoor, Mirror
Case of 12 for $121.10
Only null left
Add to Cart
Description
Description
  • Dielectric
  • Interchangeable temples and neoprene strap
  • Temples adjust to three different lengths
  • Dual frame design features a rubber gasket
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Passes D3: Droplet and Splash Test when worn with strap
  • Specifications
    Specifications
    ANSI Z87.1 ,CSA Z94.3-07
    EN-166
    41 g
    Polycarbonate
    Dielectric
    Black
    Polycarbonate
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.