Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isovaleric anhydride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.9400g/mL |
| Boiling Point | 215°C |
| Flash Point | 91°C |
| Assay Percent Range | 94% min |
| Packaging | Glass bottle |
| Quantity | 25 mL |
| Specific Gravity | 0.94 |
| Solubility Information | Solubility in water: hydrolyses |
| Formula Weight | 186.25 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Keep only in original container. |
| EINECSNumber | 215-994-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction