Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isovaleronitrile, 98%

Catalog No. p-3771085
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 625-28-5 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-28-5
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001944
InChI Key QHDRKFYEGYYIIK-UHFFFAOYSA-N
Synonym isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile
PubChem CID 12244
IUPAC Name 3-methylbutanenitrile
SMILES CC(C)CC#N

Specifications

Color Colorless
Density 0.7950g/mL
Boiling Point 128.0°C to 130.0°C (730.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CN
Quantity 5 g
Specific Gravity 0.795
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isovaleronitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF
EINECSNumber 210-884-1
Recommended Storage 4°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.