Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoviolanthrone, TCI America™
SDP

Catalog No. I022210G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 128-64-3
Molecular Formula C34H16O2
Molecular Weight (g/mol) 456.5
MDL Number MFCD00021274
InChI Key BSIHWSXXPBAGTC-UHFFFAOYSA-N
Synonym Isodibenzanthrone
PubChem CID 67189
SMILES C1=CC=C2C(=C1)C3=C4C(=CC=C5C4=C(C=C3)C6=CC=C7C8=C(C=CC5=C68)C9=CC=CC=C9C7=O)C2=O

Specifications

Color Black-Purple
Quantity 10 g
Formula Weight 456.50
Physical Form Crystalline Powder
Chemical Name or Material Isoviolanthrone
EINECSNumber (4)-0004
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.