Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISPD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560520 Supplier Invitrogen™ Supplier No. PA560520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SELNHVKVTS EALGHAGRHL QQIILDQCYN FVCVNVTTSD FQETQKLLSM LEESSLCILY PVVVVSVHFL DFKLVPPSQK MENLMQ Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 58%.

Required for protein O-linked mannosylation. Probably acts as a nucleotidyltransferase involved in synthesis of a nucleotide sugar. Required for dystroglycan O-mannosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISPD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISPD
Gene Accession No. A4D126
Gene Alias 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein; 4930579E17Rik; 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog; AV040780; Crppa; D-ribitol-5-phosphate cytidylyltransferase; hCG_1745121; hISPD; isoprenoid synthase domain containing; isoprenoid synthase domain-containing protein; Ispd; MDDGA7; MDDGC7; Nip; notch1-induced protein; testicular tissue protein Li 97
Gene Symbols ISPD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ISPD. Recombinant protein control fragment (Product #RP-100342).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.