Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ispinesib
GREENER_CHOICE

Catalog No. 540810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-081-0 10 mg
54-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-081-0 Supplier Tocris Bioscience™ Supplier No. 5408/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity and selective allosteric KSP inhibitor

Ispinesib is a high affinity allosteric kinesin spindle protein (KSP) inhibitor (Ki app = 1.7 nM). Exhibits >10,000-fold selectivity for KSP over a range of other mitotic kinesins. Inhibits proliferation and induces apoptosis of prostate and breast cancer cells in vitro. Induces tumor regression of breast cancer cell xenografts in mice.

Specifications

Quantity 10 mg
Product Type Ispinesib
Purity >98%
Molecular Weight (g/mol) 517.06

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.