Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isradipine, 98+%, PROMO, Thermo Scientific Chemicals

Catalog No. J63920MFPM
Click to view available options
Quantity:
50 mg

L-type calcium channel blocker

Applications

Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

Solubility

Soluble in DMSO

Notes

Protect from heat. Store away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5
Molecular Weight (g/mol) 371.39 g/mol
MDL Number MFCD00153820
InChI Key HMJIYCCIJYRONP-UHFFFAOYNA-N
Synonym isradipine, dynacirc, isradipin, lomir, isrodipine, prescal, dynacirc cr, esradin, clivoten, dynacrine
PubChem CID 3784
IUPAC Name 3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C

Specifications

Color Yellow
Quantity 50 mg
Merck Index 14,5240
Solubility Information Soluble in DMSO
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Isradipine
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.