Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Isradipine
GREENER_CHOICESDP

Catalog No. 200410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-041-0 10 mg
20-045-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-041-0 Supplier Tocris Bioscience™ Supplier No. 2004/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaV1.x blocker

Potent and selective L-type voltage-gated Ca2+ channel blocker. EC50 and pA2 values are 1.4 nM and 10.3 for relaxation of depolarization- and Ca2+-induced contractions of rabbit aorta respectively; EC25 = 0.45 nM for reduction in rate of spontaneously beating guinea pig right atria. Long-acting antihypertensive agent in vivo, exerting primary effects on vascular tissue with secondary negative chronotropic action.

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5

Specifications

Quantity 10 mg
Formula Weight 371.39
Purity >99%
Molecular Weight (g/mol) 371.39
Chemical Name or Material Isradipine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.