Learn More
3M™ IST AIM Logic 250 Multigas Detector
Catalog No. 19500105 Shop All 3M Products
Change view
Click to view available options
Multigas Det,logic 250
Catalog No.
19-500-105 Multigas Det,logic 250
1 options
Catalog No. 19-500-105 Supplier 3M™ Supplier No. 3250MGOXCO
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Pre-calibrated with 28 industrial gases and vapors
Calibrated to detect 28 industrial gases and vapors with dedicated LED for each sensor with an audible and visual alarm.
Features
  • Atmospheric memory
  • Programmable gas levels
  • Simple three key operation with extra large keys to accommodate gloves
  • Data can be stored and downloaded for up to 1300 intervals