Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IST1 Homolog Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS163325UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1633-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1633-25UL Supplier MilliporeSigma Supplier No. ABS163325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-IST1 Homolog, Cat. No. ABS1633,is a rabbit polyclonal antibody that detects IST1 homolog and has been tested for use in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IST1 Homolog
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias hIST1;Putative MAPK-activating protein PM28
Gene Symbols IST1;KIAA0174
Host Species Rabbit
Immunogen A recombinant fragment corresponding to the first 189 amino acids from the N-terminal half of human IST1 homolog.
Molecular Weight of Antigen ∽50 kDa observed; 39.75 kDa calculated.. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects IST1 homolog in human cells. It targets an epitope within the first 189 amino acids from the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.