Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IST1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546025
Description
Peptide sequence: INRLKLLEKK KTELAQKARK EIADYLAAGK DERARIRVEH IIREDYLVEA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Proposed to be involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis, but not for HIV-1 budding. The involvement in the MVB pathway is not established. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells.
Specifications
IST1 | |
Polyclonal | |
Unconjugated | |
IST1 | |
2400003C14Rik; AW536298; fb16b04; hIST1; hypothetical protein LOC321460; hypothetical protein LOC508900; increased sodium tolerance 1 homolog; increased sodium tolerance 1 homolog (yeast); IST1; IST1 factor associated with ESCRT-III; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; IST1, ESCRT-III associated factor; IST1-like protein; Kiaa0174; KIAA0174-like protein; MAPK activating protein PM28; MGC117220; mKIAA0174; OLC1; overexpressed in lung cancer 1; pm28; putative MAPK-activating protein PM28; RGD1307799; wu:fb16b04; wu:fi89e07; zgc:55671 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9798 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P53990 | |
IST1 | |
Synthetic peptide directed towards the N-terminal of human IST1 (aa 19-68). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction