Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559738 Supplier Invitrogen™ Supplier No. PA559738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APRLQSEVAE LKIVADQLCA KYSKEYGKLC RTNQIGTVND RLMHKLSVEA PPKILVERYL IEIAKNYNVP YEPDSVVMAE APPGVETDLI DVGFTDD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Proposed to be involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis, but not for HIV-1 budding. The involvement in the MVB pathway is not established. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IST1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IST1
Gene Accession No. P53990
Gene Alias 2400003C14Rik; AW536298; fb16b04; hIST1; hypothetical protein LOC321460; hypothetical protein LOC508900; increased sodium tolerance 1 homolog; increased sodium tolerance 1 homolog (yeast); IST1; IST1 factor associated with ESCRT-III; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; IST1, ESCRT-III associated factor; IST1-like protein; Kiaa0174; KIAA0174-like protein; MAPK activating protein PM28; MGC117220; mKIAA0174; OLC1; overexpressed in lung cancer 1; pm28; putative MAPK-activating protein PM28; RGD1307799; wu:fb16b04; wu:fi89e07; zgc:55671
Gene Symbols IST1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IST1. Recombinant protein control fragment (Product #RP-98777).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9798
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.