Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IST1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562981
Description
Immunogen sequence: LGSGFKAERL RVNLRLVINR LKLLEKKKTE LAQKARKEIA DYLAAGKDER ARIRVEHIIR EDYLVEAMEI LELYCDLL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Proposed to be involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis, but not for HIV-1 budding. The involvement in the MVB pathway is not established. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells.
Specifications
IST1 | |
Polyclonal | |
Unconjugated | |
IST1 | |
2400003C14Rik; AW536298; fb16b04; hIST1; hypothetical protein LOC321460; hypothetical protein LOC508900; increased sodium tolerance 1 homolog; increased sodium tolerance 1 homolog (yeast); IST1; IST1 factor associated with ESCRT-III; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; IST1, ESCRT-III associated factor; IST1-like protein; Kiaa0174; KIAA0174-like protein; MAPK activating protein PM28; MGC117220; mKIAA0174; OLC1; overexpressed in lung cancer 1; pm28; putative MAPK-activating protein PM28; RGD1307799; wu:fb16b04; wu:fi89e07; zgc:55671 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P53990 | |
IST1 | |
Recombinant protein corresponding to Human IST1. Recombinant protein control fragment (Product #RP-98776). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction