Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106294 Supplier Invitrogen™ Supplier No. PA5106294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IST1.

Proposed to be involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis, but not for HIV-1 budding. The involvement in the MVB pathway is not established. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IST1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IST1
Gene Accession No. P53990, Q9CX00
Gene Alias 2400003C14Rik; AW536298; fb16b04; hIST1; hypothetical protein LOC321460; hypothetical protein LOC508900; increased sodium tolerance 1 homolog; increased sodium tolerance 1 homolog (yeast); IST1; IST1 factor associated with ESCRT-III; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; IST1, ESCRT-III associated factor; IST1-like protein; Kiaa0174; KIAA0174-like protein; MAPK activating protein PM28; MGC117220; mKIAA0174; OLC1; overexpressed in lung cancer 1; pm28; putative MAPK-activating protein PM28; RGD1307799; wu:fb16b04; wu:fi89e07; zgc:55671
Gene Symbols IST1
Host Species Rabbit
Immunogen A synthesized peptide derived from human IST1(Accession P53990), corresponding to amino acid residues P246-Q296.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71955, 9798
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.