Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Istradefylline
GREENER_CHOICE

Catalog No. 514710R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-471-0R 10 mg
51-475-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-471-0R Supplier Tocris Bioscience™ Supplier No. 5147/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective adenosine A2A receptor antagonist

Potent and selective adenosine A2A receptor antagonist (Ki values are 2.2 and 150 nM for A2A and A1 receptors respectively). Anticataleptic and antiparkinson agent; reverses drug-induced motor dysfunction in animal models.

Chemical Identifiers

CAS 155270-99-8
Molecular Formula C20H24N4O4

Specifications

Quantity 10 mg
Formula Weight 384.43
Physical Form Solid
Color White
Purity >99%
Molecular Weight (g/mol) 384.44
Chemical Name or Material Istradefylline
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.