Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISX/Intestine specific homeobox Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984181
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4181 Supplier Bioss Supplier No. BS18172R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the RAXLX homeobox gene family.Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the RAXLX homeobox gene family.

Specifications

Antigen ISX/Intestine specific homeobox
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ISX
Gene Accession No. Q2M1V0
Gene Alias DKFZp781N2395; intestine specific homeobox; Intestine-specific homeobox; Intestine-specific homeobox-like; ISX; LOC100351992; MGC138417; pancreas-intestine homeodomain transcription factor; Pix-1; RAX-like homeobox; RAXLX
Gene Symbols ISX
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ISX/Intestine specific homeobox.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91464
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.