Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543052 Supplier Invitrogen™ Supplier No. PA543052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ILKRPARRSD MDRPEGPGEE GPGEAAASGS GLEKPPKDQP QEGRKSKRRV Sequence homology: Human: 100%; Rabbit: 83%; Rat: 77%.

Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the RAXLX homeobox gene family.Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the RAXLX homeobox gene family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISX
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ISX
Gene Accession No. Q2M1V0
Gene Alias 9130012O13Rik; DKFZp781N2395; intestine specific homeobox; intestine-specific homeobox; ISX; MGC138417; pancreas-intestine homeodomain transcription factor; Pix-1; RAX-like homeobox; RAXLX
Gene Symbols ISX
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ISX (aa 36-85).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91464
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.