Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553529 Supplier Invitrogen™ Supplier No. PA553529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQEYEKKLRA ELVEKWKAER EARLARGEKE EEEEEEEEIN IYAVTEEESD EEGSQEKGGD DSQQKFIAHV PVPSQQEIEE ALVRRKKMEL LQKYASETLQ AQSEEARR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

ISY1 may play a role in pre-mRNA splicing as component of the spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISY1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISY1
Gene Accession No. Q9ULR0
Gene Alias 5830446M03Rik; AI181014; AU020769; FSAP33; functional spliceosome-associated protein 33; ISY1; ISY1 splicing factor homolog; ISY1 splicing factor homolog (S. cerevisiae); Isy1-rab43; ISY1-RAB43 readthrough; KIAA1160; Pre-mRNA-splicing factor ISY1 homolog; RGD1307688
Gene Symbols ISY1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ISY1. Recombinant protein control fragment (Product #RP-89213).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57461
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.