Learn More
Invitrogen™ ISY1 Polyclonal Antibody

Description
Immunogen sequence: MARNAEKAMT ALARFRQAQL EEGKVKERRP FLASECTELP KAEKWRRQII GEISKKVAQI QNAGLGEFRI RDLNDEINKL LREKGHWEVR IKELGGPDYG KVGPKMLDHE GKEVPGNRGY KYFGAAKDLP GVRELFEKER QVRWLMPVIP ALWEAEAGGS QALPPPRKTR AELMKAIDFE YYGYLDEDDG VIVPLEQEYE KKLRAELVEK WKAEREARLA RGEKEEEEEE EEEINIYAVT EEESDEEGSQ EKGGDDSQQK FIAHVPVPSQ QEIEEALVRR KKMELLQKYA SETLQAQSEE ARRLLGY.
Specifications
Specifications
| Antigen | ISY1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.64 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ISY1 |
| Gene Accession No. | Q69ZQ2, Q9ULR0 |
| Gene Alias | 5830446M03Rik; AI181014; AU020769; FSAP33; functional spliceosome-associated protein 33; ISY1; ISY1 splicing factor homolog; ISY1 splicing factor homolog (S. cerevisiae); Isy1-rab43; ISY1-RAB43 readthrough; KIAA1160; Pre-mRNA-splicing factor ISY1 homolog; RGD1307688 |
| Gene Symbols | ISY1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.