Learn More
ISY1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304927A

Description
The recommended shelf life for this product is 1 year from date of receipt.
ISY1 may play a role in pre-mRNA splicing as component of the spliceosome.Specifications
ISY1 | |
Polyclonal | |
Unconjugated | |
ISY1 | |
fSAP33, functional spliceosome-associated protein 33, ISY1 splicing factor homolog (S. cerevisiae), KIAA1160 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57461 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9ULR0 | |
ISY1 | |
Between 200 and 250 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.