Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISYNA1 Polyclonal Antibody
ISYNA1 Polyclonal Antibody
Supplier: Thermo Scientific PA5100272
Description
ISYNA1 Polyclonal Antibody for Western Blot

Specifications
ISYNA1 | |
Polyclonal | |
Unconjugated | |
ISYNA1 | |
1300017C10Rik; AU018670; hINO1; Ino1; INOS; Inositol-3-phosphate synthase 1; IPS; IPS 1; IPS-1; Isyna1; MGC133800 protein; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; myo-inositol-1-phosphate synthase; testis secretory sperm-binding protein Li 200a | |
Rabbit | |
Affinity Chromatography | |
RUO | |
290651, 51477, 71780 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6AYK3, Q9JHU9, Q9NPH2 | |
ISYNA1 | |
A synthesized peptide derived from human ISYNA1(Accession Q9NPH2), corresponding to amino acid residues L515-T558. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction