Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISYNA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552487 Supplier Invitrogen™ Supplier No. PA552487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVLVDFLIGS GLKTMSIVSY NHLGNNDGEN LSAPLQFRSK EVSKSNVVDD MVQSNPVLYT PGEEPDHCVV IKYVPYVGDS KRALDEYTSE LMLGGTNTLV LHNTCEDSLL AAPIMLDLA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate. Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate (Seelan et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISYNA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISYNA1
Gene Accession No. Q9NPH2
Gene Alias 1300017C10Rik; AU018670; hINO1; Ino1; INOS; Inositol-3-phosphate synthase 1; IPS; IPS 1; IPS-1; Isyna1; MGC133800 protein; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; myo-inositol-1-phosphate synthase; testis secretory sperm-binding protein Li 200a
Gene Symbols ISYNA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ISYNA1. Recombinant protein control fragment (Product #RP-89548).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51477
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.