Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itaconic acid, 99+%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
97-65-4
C5H6O4
130.1
LVHBHZANLOWSRM-UHFFFAOYSA-N
itaconic acid,2-methylenesuccinic acid,methylenesuccinic acid,methylenebutanedioic acid,propylenedicarboxylic acid,itaconate,2-propene-1,2-dicarboxylic acid,butanedioic acid, methylene,succinic acid, methylene,2-methylenebutanedioic acid
811
CHEBI:30838
2-methylidenebutanedioic acid
C=C(CC(=O)O)C(=O)O
97-65-4
C5H6O4
130.1
LVHBHZANLOWSRM-UHFFFAOYSA-N
itaconic acid,2-methylenesuccinic acid,methylenesuccinic acid,methylenebutanedioic acid,propylenedicarboxylic acid,itaconate,2-propene-1,2-dicarboxylic acid,butanedioic acid, methylene,succinic acid, methylene,2-methylenebutanedioic acid
811
CHEBI:30838
2-methylidenebutanedioic acid
C=C(CC(=O)O)C(=O)O
Infrared Spectrum Authentic
Sulfated Ash 0.1% max.
Density 1.5700g/mL
Water 0.5% max.
Packaging Plastic drum
Specific Gravity 1.57
Quantity 5kg
Color White to Beige
Melting Point 165.0°C to 168.0°C
Assay Percent Range 99+%
Linear Formula HO2CCH2C(=CH2)CO2H
Beilstein 02,76
Merck Index 15,529
Solubility Information Solubility in water: 95g/L (25°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in most other common organic,solvents
Formula Weight 130.1
Physical Form Crystalline Powder or Crystals
Percent Purity ≥99%
Chemical Name or Material Itaconic acid
Show More Show Less