Learn More
Itaconic anhydride, 97%
Description
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.

Specifications
Specifications
Melting Point | 68°C to 72°C |
Boiling Point | 114°C to 115°C (12 mmHg) |
Quantity | 500 g |
Beilstein | 2212 |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 112.09 |
Percent Purity | 97% |
Chemical Name or Material | Itaconic anhydride |
Safety and Handling
Hazard Category | H318 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P280-P305+P351+P338-P310 |
EINECSNumber | 218-518-2 |
TSCA | Yes |
Recommended Storage | Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.