Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116631 Supplier Invitrogen™ Supplier No. PA5116631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, THP-1, NIH/3T3, C6, Mouse lung, Mouse kidney, Rat lung, Rat spleen Immunogen sequence: MSDSGSQLGS MGSLTMKSQL QITVISAKLK ENKKNWFGPS PYVEVTVDGQ SKKTEKCNNT NSPKWKQPLT VIVTPVSKLH FRVWSHQTLK SDVLLGTAAL DIYETLKSNN MKLEEVVVTL QLGGDKEPTE TIGDLSICLD GLQLESEVVT NGETT.

Atrophin-1 contains a polyglutamine repeat, expansion of which is responsible for dentatorubral and pallidoluysian atrophy. The protein encoded by this gene interacts with atrophin-1. This encoded protein is a closely related member of the NEDD4-like protein family. This family of proteins are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. This encoded protein contains four tandem WW domains and a HECT domain. It can act as a transcriptional corepressor of p45/NFE2 and may participate in the regulation of immune responses by modifying Notch-mediated signaling. It is highly similar to the mouse Itch protein, which has been implicated in the regulation and differentiation of erythroid and lymphoid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITCH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ITCH
Gene Accession No. Q8C863, Q96J02
Gene Alias 6720481N21Rik; 8030492O04Rik; A130065M08; ADMFD; AIF4; AIP4; atrophin-1 interacting protein 4; Atrophin-1-interacting protein 4; C230047C07Rik; dJ468O1.1; E3 ubiquitin-protein ligase Itchy; E3 ubiquitin-protein ligase Itchy homolog; HECT-type E3 ubiquitin transferase Itchy homolog; ITCH; itch E3 ubiquitin ligase; itchy E3 ubiquitin protein ligase; itchy E3 ubiquitin protein ligase homolog; itchy, E3 ubiquitin protein ligase; NAPP1; NFE2-associated polypeptide 1
Gene Symbols ITCH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-155 of ITCH (NP_0012440661).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16396, 311567, 83737
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.