Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ITD 1

Catalog No. 506850R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-685-0R 50 mg
50-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-685-0R Supplier Tocris Bioscience™ Supplier No. 5068/50
This item is not returnable. View return policy

Selective inhibitor of TGF-β signaling; induces cardiomyocyte differentiation in ESCs

ITD 1 is a selective inhibitor of TGF-β signaling (IC50 = 0.85 μM); displays little or no inhibition of activin, Wnt or BMP signaling pathways. Induces proteasomal degradation of the TGF-β type II receptor. Inhibits TGF-β-induced mesoderm formation from mouse embryonic stem cells (ESCs) during early differentiation; selectively promotes the differentiation of ESCs to cardiomyocytes in vitro between days 3-5. Does not induce differentiation of vascular smooth muscle cells or endothelial cells.

Specifications

Purity >98%
Content And Storage -20°C
Quantity 50 mg
Product Type ITD 1
Molecular Weight (g/mol) 415.52
Inhibitors ITD 1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.