Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITFG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144724
Description
Immunogen sequence: VSFKNHSALI TSVVPGDYDG DSQMDVLLTY LPKNYAKSEL GAVIFWGQNQ TLDPNNMTIL NRTFQDEPLI MDFNGDLIPD IFGITNESNQ PQILLGGNLS WHPALTTTSK MRIPHSHAFI DLTEDFTA Antigen sequence identity: Mouse 87%; Rat 86%.
Modulator of T-cell function. Has a protective effect in graft versus host disease model.
Specifications
ITFG1 | |
Polyclonal | |
Unconjugated | |
Itfg1 | |
2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP | |
Rabbit | |
Affinity chromatography | |
RUO | |
81533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TB96 | |
Itfg1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 91-218. | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction