Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITFG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553328 Supplier Invitrogen™ Supplier No. PA553328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NCQKSTIYLV RSGMKQWVPV LQDFSNKGTL WGFVPFVDEQ QPTEIPIPIT LHIGDYNMDG YPDALVILKN TSGSNQQAFL LENVPCNNAS CEEARRMFKV YWELTDLNQI KDAMVATFFD IYEDGILDIV VLSKGYTKND Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

Modulator of T-cell function. Has a protective effect in graft versus host disease model.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITFG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Itfg1
Gene Accession No. Q8TB96
Gene Alias 2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP
Gene Symbols Itfg1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ITFG1. Recombinant protein control fragment (Product #RP-90953).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.