Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITFG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590805 Supplier Invitrogen™ Supplier No. PA590805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWGFVPFVDE QQPTEIPIPI TLHIGDYNMD GYPDALVILK NTSGSNQQAF LLENVPCNNA SCEEARRMFK VYWELTDLNQ IKDAMVATFF DIYEDGILDI VVLSKGYTKN DFAIHTLKNN FEADAYFVKV IVLSGLCSND CPRKITPFGV NQPGPYIMYT TVDANGYLKN GSAGQLSQSA HLALQLPYNV LGLGRSANFL DHLYVGIPRP SGEKSIRKQE WTAIIPNSQL IVIPYPHNVP RSWSAKLYLT PSNIVLLTAI ALIGVCVFIL AIIGILHWQE KKADDREKRQ EAHRFHFDAM; Positive Samples: LO2, A-431, Mouse heart, Mouse pancreas, Mouse testis, ; Cellular Location: Membrane, Single-pass type I membrane protein.

Modulator of T-cell function. Has a protective effect in graft versus host disease model.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITFG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Itfg1
Gene Accession No. Q8R4E1, Q8TB96, Q99KW9
Gene Alias 2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP
Gene Symbols Itfg1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171083, 71927, 81533
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.