Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITFG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | ITFG1 |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119209
|
Invitrogen™
PA5119209 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121819
|
Invitrogen™
PA5121819 |
20 μL |
Each for $178.40
|
|
|||||
Description
ITFG1 Polyclonal Antibody for Western Blot, IHC (P)
Modulator of T-cell function. Has a protective effect in graft versus host disease model.Specifications
ITFG1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Itfg1 | |
2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP | |
Itfg1 | |
Primary | |
Antigen affinity chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8TB96 | |
81533 | |
Recombinant protein Integrin Alpha FG GAP Repeat Containing Protein 1. The antigen corresponds to amino acid range 55-306 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title