Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITFG2 Monoclonal Antibody (OTI3A2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503051

Description
As part of the KICSTOR complex functions in the amino acid-sensing branch of the TORC1 signaling pathway. Recruits, in an amino acid-independent manner, the GATOR1 complex to the lysosomal membranes and allows its interaction with GATOR2 and the RAG GTPases. Functions upstream of the RAG GTPases and is required to negatively regulate mTORC1 signaling in absence of amino acids. In absence of the KICSTOR complex mTORC1 is constitutively localized to the lysosome and activated. The KICSTOR complex is also probably involved in the regulation of mTORC1 by glucose.Specifications
ITFG2 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q969R8 | |
ITFG2 | |
Full length human recombit protein of human ITFG2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI3A2 | |
Unconjugated | |
ITFG2 | |
FGGAP1, MDS028 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55846 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction