Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITFG3 Antibody (OTI3B3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272253FR
Description
ITFG3 Monoclonal specifically detects ITFG3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ITFG3 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ITFG3 | |
Full length recombinant protein of human ITFG3 (NP_114428) produced in HEK293T cells. | |
0.1 mL | |
83986.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3B3 | |
Western Blot, Immunohistochemistry | |
C16orf9, chromosome 16 open reading frame 9, DKFZP761D0211, FLJ32603, gs19, integrin alpha FG-GAP repeat containing 3, protein ITFG3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction