Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITFG3 Monoclonal Antibody (A5-A10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA542564
This item is not returnable.
View return policy
Description
Positive Control: MCF-7 cell lysate, Siha cell lysate, SHSY5Y cell lysate, LOVO. Subcellular Location: Membrane.
ITFG3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cell surface; extracellular exosome; integral component of membrane.
Specifications
ITFG3 | |
Monoclonal | |
2 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9H0X4 | |
FAM234A | |
Recombinant protein within human 300-552. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
A5-A10 | |
Unconjugated | |
FAM234A | |
C16orf9; Fam234a; family with sequence similarity 234 member A; family with sequence similarity 234, member A; gs19; integrin alpha FG-GAP repeat containing 3; ITFG3; Protein FAM234A; protein ITFG3; RGD1311484 | |
Mouse | |
Protein G | |
RUO | |
83986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction