Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITFG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584880 Supplier Invitrogen™ Supplier No. PA584880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REEVSGHLYS GSTGHQIGLR GSLGVDGESG FLLHVTRTGA HYILFPCASS LCGCSVKGLY EKVTGSGGPF KSDPHWESML NATTRRM Highest antigen sequence indentity to the following orthologs: Rat - 27%, Mouse - 69%.

ITFG3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cell surface; extracellular exosome; integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITFG3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM234A
Gene Accession No. Q9H0X4
Gene Alias C16orf9; Fam234a; family with sequence similarity 234 member A; family with sequence similarity 234, member A; gs19; integrin alpha FG-GAP repeat containing 3; ITFG3; Protein FAM234A; protein ITFG3; RGD1311484
Gene Symbols FAM234A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM234A. Recombinant protein control fragment (Product #RP-108590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.