Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITGA10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567829
Description
Predicted to react with Mouse samples.
Integrin alpha-10/beta-1 is a receptor for collagen.
Specifications
ITGA10 | |
Polyclonal | |
Unconjugated | |
ITGA10 | |
A630048L14; integrin alpha 10; integrin alpha-10; integrin subunit alpha 10; integrin, alpha 10; ITGA10; PRO827; UNQ468/PRO827 | |
Rabbit | |
affinity chromatography | |
RUO | |
213119, 8515 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75578 | |
ITGA10 | |
Synthesized peptide derived from human ITGA10. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction