Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100840 Supplier Invitrogen™ Supplier No. PA5100840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ITGA10.

Integrin alpha-10/beta-1 is a receptor for collagen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ITGA10
Gene Accession No. O75578
Gene Alias A630048L14; integrin alpha 10; integrin alpha-10; integrin subunit alpha 10; integrin, alpha 10; ITGA10; PRO827; UNQ468/PRO827
Gene Symbols ITGA10
Host Species Rabbit
Immunogen A synthesized peptide derived from human ITGA10(Accession O75578), corresponding to amino acid residues P213-G263.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213119, 8515
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.