Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA11 Monoclonal Antibody (1804CT459.14.18)
GREENER_CHOICE

Catalog No. PIMA524758
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524758 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524758 Supplier Invitrogen™ Supplier No. MA524758
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ITGA11 Monoclonal Antibody for Western Blot

This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein contains an I domain, is expressed in muscle tissue, dimerizes with beta 1 integrin in vitro, and appears to bind collagen in this form. Therefore, the protein may be involved in attaching muscle tissue to the extracellular matrix. Alternative transcriptional splice variants have been found for this gene, but their biological validity is not determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA11
Applications Western Blot
Classification Monoclonal
Clone 1804CT459.14.18
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Itga11
Gene Accession No. Q9UKX5
Gene Alias 4732459H24Rik; HsT18964; integrin a11; integrin alpha 11; integrin alpha-11; integrin subunit alpha 11; integrin, alpha 11; Itga11; MSTP018
Gene Symbols Itga11
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 675-880 amino acids from human ITGA11.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.