Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITGA2 Monoclonal Antibody (AK7), Biotin
GREENER_CHOICE

Catalog No. PIMA180244
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA180244 Supplier Invitrogen™ Supplier No. MA180244
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD49b antibody, clone AK7 recognizes the integrin alpha 2 subunit, a approximately 160 kDa glycoprotein that non-covalently associates with the approximately 130 kDa integrin beta 1 subunit to form the VLA-2 complex.

CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGA2
Applications Flow Cytometry
Classification Monoclonal
Clone AK7
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene Itga2
Gene Accession No. P17301
Gene Alias alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2
Gene Symbols Itga2
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3673
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.