Learn More
Invitrogen™ ITGA2 Monoclonal Antibody (DX5), Biotin

Description
ITGA2 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | ITGA2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DX5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | Itga2 |
| Gene Accession No. | Q62469 |
| Gene Alias | alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.