Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA2 Mouse anti-Human, Clone: TEA1/41 (FITC), Abnova™

Catalog No. 89033753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-753 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-753 Supplier Abnova Corporation Supplier No. MAB13688
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human ITGA2.

  • High specificity
  • High reactivity

Specifications

Antigen ITGA2
Applications Flow Cytometry
Classification Monoclonal
Clone TEA1/41
Conjugate FITC
Description Mouse monoclonal antibody raised against human ITGA2.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Gene Accession No. P17301
Gene Alias BR/CD49B/GPIa/VLA-2/VLAA2
Gene Symbols ITGA2
Host Species Mouse
Immunogen TNF activated HUVEC cells.
Purification Method Protein A/G purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3673
Target Species Human
Content And Storage Store in the dark at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.