Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITGA2B Mouse anti-Human, Clone: HIP8 (FITC), Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13899
Description
Mouse monoclonal antibody raised against human ITGA2B.
- High specificity
- High reactivity
Specifications
ITGA2B | |
Monoclonal | |
FITC | |
Flow Cytometry (20 uL/106 cells) Immunohistochemistry (Frozen sections) The optimal working dilution should be determined by the end user. | |
P08514 | |
ITGA2B | |
Idiopathic thrombocytopenic purpura (IPT) patient platelets. | |
100 Reactions | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
HIP8 | |
Mouse monoclonal antibody raised against human ITGA2B. | |
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
CD41/CD41B/GP2B/GPIIb/GTA/HPA3 | |
Mouse | |
Protein A/G purification | |
RUO | |
3674 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction